missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human beta-Defensin 1 (47 aa) Recombinant Protein, PeproTech®

Product Code. 17812643
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17812643

Brand: Gibco™ 30051A100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BD-1 is a 5.0 kDa protein containing 47 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60022
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1672
Molecular Weight (g/mol) 5 kDa
Name Human beta-Defensin 1 (47 aa)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1
Common Name beta-Defensin 1
Gene Symbol DEFB1
Biological Activity Determined by its ability to chemoattract CD34+ dendritic cells using a concentration range of 100.0-1000.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GNFLTGLGHR SDHYNCVSSG GQCLYSACPI FTKIQGTCYR GKAKCCK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.