missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-Catenin Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB13291-050
Additional Details : Weight : 0.00970kg
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
beta-Catenin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1499 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
196618 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CTNNB1 | |
E. coli-derived recombinant human beta-Catenin aa 2-781 | |
50 μg | |
Primary | |
Detects human beta-Catenin in direct ELISAs. | |
Human | |
Purified |