missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta 2-Microglobulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB82481-100
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| beta2-Microglobulin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| B2M, beta 2Microglobulin, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Chinese hamster ovary cell line CHO-derived recombinant human beta 2 -Microglobulin Met1-Met119 Accession # P61769 | |
| 100 μg | |
| 567 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| CyTOF, Flow Cytometry, Western Blot | |
| 883005 | |
| Western Blot 2μg/mL, Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| P61769 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human beta 2 -Microglobulin in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction