missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-1,4-Galactosyltransferase 1/B4GalT1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£186.00 - £490.00
Specifications
| Antigen | beta-1,4-Galactosyltransferase 1/B4GalT1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18739057
![]() |
R&D Systems
AF3609-SP |
25 μg |
£186.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003392
|
R&D Systems
AF3609 |
100 μg |
£490.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1,4-Galactosyltransferase 1/B4GalT1 Polyclonal specifically detects beta-1,4-Galactosyltransferase 1/B4GalT1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| beta-1,4-Galactosyltransferase 1/B4GalT1 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| 1,4- galactosyltransferase, polypeptide 1, B4GAL-T1, beta-1,4-galactosyltransferase 1, Beta-1,4-GalTase 1, Beta4Gal-T1, betaGlcNAc beta, CDG2D, EC 2.4.1, GGTB2, GGTB2DKFZp686N19253, glycoprotein-4-beta-galactosyltransferase 2, GT1, GTB, lactose synthase, MGC50983, UDP-Gal, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1 | |
| B4GALT1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P15291 | |
| 2683 | |
| Mouse myeloma cell line NS0-derived recombinant human beta-1,4-Galactosyltransferase 1/B4GalT1 long isoform Gly44-Ser398 Accession # P15291 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human beta-1,4-Galactosyltransferase 1/B4GalT1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title