missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8560-100
This item is not returnable.
View return policy
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), CD57, EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATP, GLCATPGLCUATP, glcUAT-P, GLCUATP, Glucuronosyltransferase P, HNK1, LEU7, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| Chinese Hamster Ovary cell line, CHO-derived human beta-1,3-Glucuronyltransferase 1/B3GAT1, His25-Ile334, Accession # Q9P2W7 | |
| 100 μg | |
| 27087 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 1002707 | |
| Immunohistochemistry 5-25 μg/mL | |
| Q9P2W7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human beta-1,3-Glucuronyltransferase 1/B3GAT1 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction