missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human BERP Control Fragment Recombinant Protein

Produktkod. 30181472
Klicka för att se tillgängliga alternativ
Quantity:
100 μL
Förpackningsstorlek:
100µL
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 30181472

Brand: Invitrogen™ RP98385

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83656 (PA5-83656. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the tripartite motif family, also called the 'RING-B-box-coiled-coil' subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic filaments. It is similar to a rat protein which is a specific partner for the tail domain of myosin V, a class of myosins which are involved in the targeted transport of organelles. The rat protein can also interact with alpha-actinin-4. Thus it is suggested that this human protein may play a role in myosin V-mediated cargo transport. Alternatively spliced transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number O75382
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10612
Name Human BERP Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Berp; BERP1; brain expressed ring finger; brain-expressed RING finger protein; Hac1; RING finger protein 22; RING finger protein 97; RING finger protein HAC1; RNF22; RNF97; Trim3; tripartite motif containing 3; tripartite motif protein 3; tripartite motif protein TRIM3; tripartite motif-containing 3; tripartite motif-containing protein 3
Common Name BERP
Gene Symbol TRIM3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TQRQGQEHIGSSCSFAEQALRLGSAPEVLLVRKHMRERLAALAAQAFPERPHENAQLELVLEVDGLRRSVLNLGALLTTSATAHETVAT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.