missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF Antibody, R&D Systems™

Chicken Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF248
This item is not returnable.
View return policy
Description
BDNF Polyclonal specifically detects BDNF in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| BDNF | |
| Polyclonal | |
| Unconjugated | |
| P23560 | |
| BDNF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF His129-Arg247 Accession # P23560 | |
| 100 μg | |
| Primary | |
| Detects human BDNF in direct ELISAs. | |
| Human | |
| IgY |
| Flow Cytometry, CyTOF | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 μg/106 cells, CyTOF-ready | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Chicken | |
| Antigen Affinity-purified from egg yolks | |
| RUO | |
| 627 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction