missing translation for 'onlineSavingsMsg'
Learn More

Human BDNF Antibody, R&D Systems™

Product Code. 30713195
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713195 100 μg 100µg
30713196 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30713195 Supplier R&D Systems Supplier No. MAB248R100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for Western Blot,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDNF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 35928R
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. P23560
Gene Alias abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF, Arg128-Arg247, Accession # P23560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 627
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.