missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-5, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
Description
Recombinant Human BD-5 is a 5.8 kDa protein containing 51 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
Specifications
Specifications
| Accession Number | Q8NG35 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 245908, 504180 |
| Molecular Weight (g/mol) | 5.8 kDa |
| Name | Human BD-5, Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 5 μg |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction