missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BCMA/TNFRSF17 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1931N-100UG
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.
Specifications
| BCMA/TNFRSF17 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
| Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17, Met1-Ala54, Accession No. Q02223 | |
| Primary | |
| Detects human BCMA/TNFRSF17 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
| Flow Cytometry | |
| 1004023 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q02223 | |
| Mouse | |
| 100 μg | |
| 608 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction