missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bcl9-2 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4967
This item is not returnable.
View return policy
Description
BCL9-2 Polyclonal specifically detects BCL9-2 in Human samples. It is validated for Western Blot.
Specifications
| BCL9-2 | |
| Polyclonal | |
| Unconjugated | |
| Q86UU0 | |
| BCL9L | |
| E. coli-derived recombinant human Bcl9-2 Met38-Val206 Accession # Q86UU0 | |
| 100 μg | |
| Primary | |
| Detects endogenous human Bcl9-2 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, Bcl92, BCL9-like protein, DLNB11, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 283149 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction