missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Bcl-X (aa 1-70) Control Fragment Recombinant Protein

Product Code. 30200389
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30200389 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30200389 Supplier Invitrogen™ Supplier No. RP102677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded the BCL-X gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The proteins encoded by this gene are located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential, and thus controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Alternative splicing results in multiple transcript variants encoding two different isoforms. The longer isoform acts as an apoptotic inhibitor and the shorter isoform acts as an apoptotic activator.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07817
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 598
Name Human Bcl-X (aa 1-70) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias anti-apoptosis regulatory protein; anti-apoptotic Bcl-2 gene family member; anti-apoptotic Bcl-xL; anti-apoptotic regulator Bcl-xL; apoptosis regulator Bcl-X; B cell lymphoma 2 like; B cell lymphoma like X; B-cell leukemia/lymphoma x; Bcl 2 like 1; bcl x; Bcl xL; BCL XL/S; Bcl(X)L; bcl-2 L1; BCL2 like 1; BCL2L; Bcl2l1; bcl2-L-1; BCL2-like 1; Bcl-2-like protein 1; BCL2-like protein 1; BCLX; bcl-x; Bcl-x long protein; BclxL; Bcl-xL; BCL-XL/S; BclXS; bcl-xS; BLC2L; DKFZp781P2092; PPP1R52; protein phosphatase 1, regulatory subunit 52; RP5-857M17.3; unnamed protein product
Common Name BCL-X
Gene Symbol BCL2L1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MSQSNRELVVDFLSYKLSQKGYSWSQFSDVEENRTEAPEGTESEMETPSAINGNPSWHLADSPAVNGATG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.