missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bcl-w Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB824-SP
This item is not returnable.
View return policy
Description
BCL-W/BCL2L2 Monoclonal specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot.
Specifications
| BCL-W/BCL2L2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q92843 | |
| BCL2L2 | |
| E. coli-derived recombinant human Bcl-w Met1-Thr172 Accession # Q92843 | |
| 25 μg | |
| Primary | |
| Detects human Bcl-w in Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG1 |
| Western Blot | |
| 90316 | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS with Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2L2, BCL2-like 2, Bclw, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 599 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction