missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Bcl-B (aa 1-33) Control Fragment Recombinant Protein

Product Code. 30182172
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182172

Brand: Invitrogen™ RP98457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111149 (PA5-111149. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HD36
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10017
Name Human Bcl-B (aa 1-33) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA420380; anti-apoptotic protein Boo; Anti-apoptotic protein NrH; apoptosis regulator Bcl-B; AU023065; Bcl-2 homolog Diva; BCL2 like 10; BCL2L10; bcl2-L-10; Bcl2-like 10; BCL2-like 10 (apoptosis facilitator); bcl-2-like protein 10; BCLB; BCL-B; Boo; C85687; death inducer binding to vBcl-2 and Apaf-1; Diva
Common Name Bcl-B
Gene Symbol BCL2L10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MVDQLRERTTMADPLRERTELLLADYLGYCARE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.