missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bcl-2 Minus C-Terminus Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £361.00
Specifications
| Antigen | Bcl-2 |
|---|---|
| Clone | 118712 |
| Dilution | Neutralization |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733318
![]() |
R&D Systems
MAB8271-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18112871
![]() |
R&D Systems
MAB8271 |
100 μg |
£361.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human samples. It is validated for Neutralization.Specifications
| Bcl-2 | |
| Neutralization | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 596 | |
| E. coli-derived recombinant human Bcl-2 minus the carboxyl-terminal mitochondrial targeting sequence | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Bcl-2 Minus C-Terminus. Neutralizes recombinant human (rh) Bcl-2 bioactivity but not rhBcl-w or rhBcl-x bioactivity. |
| 118712 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl2, Bcl-2 | |
| BCL2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title