missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BCAM Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB148
Additional Details : Weight : 0.00970kg
Description
BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for Western Blot.Specifications
BCAM/CD239 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P50895 | |
BCAM | |
Mouse myeloma cell line NS0-derived recombinant human BCAM Glu32-Ala547 Accession # P50895 | |
500 μg | |
Primary | |
Detects human BCAM in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ALCAM, rhEpCAM, recombinant mouse (rm) MAdCAM, rhMCAM, rhNCAM-L1, rmOCAM, or rmTROP-2 is observed. | |
Human | |
Purified |
Western Blot | |
87202 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4059 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only