missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bax Minus C-Terminus Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB846
This item is not returnable.
View return policy
Description
Bax Monoclonal specifically detects Bax in Human samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.
Specifications
| Bax | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
| Q07812 | |
| BAX | |
| E. coli-derived recombinant human Bax aa 1-171 Accession # Q07812 | |
| 100 μg | |
| Primary | |
| Detects full-length human Bax in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, KnockDown | |
| 127606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 581 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction