missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BATF3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7437-SP
This item is not returnable.
View return policy
Description
BATF3 Polyclonal specifically detects BATF3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| BATF3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55509 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9NR55 | |
| BATF3 | |
| E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55 | |
| 25 μg | |
| Primary | |
| Detects human BATF3 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse BATF3 is observed, and less than 2% cross-reactivity with recombinant human BATF is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering