missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BATF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7437
This item is not returnable.
View return policy
Description
BATF3 Monoclonal specifically detects BATF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| BATF3 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human BATF3 in ELISA. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 841702 | |
| Unconjugated | |
| Q9NR55 | |
| BATF3 | |
| E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55 | |
| 100 μg | |
| 55509 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction