Learn More
Human BATF Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB8054
Product Code. 18171921
  • £361.00 / 100µg
Description
BATF Monoclonal specifically detects BATF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BATF
Monoclonal
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human BATF in ELISA and Western Blot.
Human
Purified
Western Blot, Flow Cytometry, CyTOF
687706
Unconjugated
Q16520
BATF
E. coli-derived recombinant human BATF Met1-Pro125 Accession # Q16520
100 μg
10538
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b