missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BAP3 (aa 776-915) Control Fragment Recombinant Protein

Product Code. 30208249
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208249

Brand: Invitrogen™ RP90656

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53347 (PA5-53347. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BAP3 was initially identified through interaction in a yeast two-hybrid system with the brain-specific angiogenesis inhibitor 1, a p53-target gene that encodes a seven-span transmembrane protein member of the secretin receptor family. BAP3 is predominantly expressed in the brain and possess high homology with Munc13 and synaptotagmin, suggesting that BAP3 may play a role in regulating neurotransmitter release. Recent experiments have shown that BAP3 is induced in certain tumors such as desmoplastic small round cell tumor. Ectopic expression of BAP3 in tumor cells dramatically enhances growth in low serum conditions and colony formation in soft agar, suggesting that the regulated exocytotic pathway may play a role in cancer cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94812
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8938
Name Human BAP3 (aa 776-915) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AL031709.1; BAI1 associated protein 3; BAI1-associated protein 3; BAIAP3; BAI-associated protein 3; BAP3; Brain-specific angiogenesis inhibitor I-associated protein 3; Gm937; KIAA0734
Common Name BAP3
Gene Symbol BAIAP3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ATLFYTELLRKKVDTQPGAAGEAVSEALCVVLNNVELVRKAAGQALKGLAWPEGATGPEGVLPRPLLSCTQALDDDLQREAHTVTAHLTSKMVGDIRKYVQHISLSPDSIQNDEAVAPLMKYLDEKLALLNASLVKGNLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.