missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BANP Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1144

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15225149

  • £112.00 / 50µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BANP Synthetic Peptide, PEP-1144, from Invitrogen.

This peptide corresponds to 14 amino acids near the center of human BANP.PEP-1144 can be used as a blocking peptide with polyclonal antibody PA5-21030.
Specifications

Specifications

54971
purified
Human BANP
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q8N9N5
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
BANP
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers