missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bag-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB852-SP
This item is not returnable.
View return policy
Description
Bag-1 Monoclonal specifically detects Bag-1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Knockout Validated.
Specifications
| Bag-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunoprecipitation 0.6 ug/10^6 cells, Knockout Validated | |
| BAG family molecular chaperone regulator 1, Bag1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 573 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunoprecipitation, Immunoassay | |
| GP3.10G3E2 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAG1 | |
| E. coli-derived recombinant human Bag-1 | |
| 25 μg | |
| Primary | |
| Detects the 36ákDa and 50ákDa isoforms of human Bag-1. Does not detect mouse Bag-1. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction