missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BABAM1 (aa 16-99) Control Fragment Recombinant Protein

Product Code. 30203280
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203280

Brand: Invitrogen™ RP104996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65868 (PA5-65868. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

HSPC142 is a component of the BRCA1-A complex, a complex that specifically recognizes 'Lys-63'-linked ubiquitinated histones H2A and H2AX at DNA lesions sites, leading to target the BRCA1-BARD1 heterodimer to sites of DNA damage at double-strand breaks (DSBs). The BRCA1-A complex also possesses deubiquitinase activity that specifically removes 'Lys-63'-linked ubiquitin on histones H2A and H2AX. In the BRCA1-A complex, it is required for the complex integrity and its localization at DSBs. HSPC142 probably also plays a role as a component of the BRISC complex, a multiprotein complex that specifically cleaves 'Lys-63'-linked ubiquitin. In these 2 complexes, it is probably required to maintain the stability of BRE/BRCC45 and help the 'Lys-63'-linked deubiquitinase activity mediated by BRCC3/BRCC36.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NWV8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29086
Name Human BABAM1 (aa 16-99) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5430437P03Rik; BABAM1; BRCA1-A complex subunit MERIT40; BRISC and BRCA1 A complex member 1; BRISC and BRCA1-A complex member 1; C19orf62; HSPC142; mediator of Rap80 interactions and targeting 40 kDa; mediator of RAP80 interactions and targeting subunit of 40 kDa; MERIT40; NBA1; new component of the BRCA1 A complex; new component of the BRCA1-A complex; new component of the BRCAA1 A complex
Common Name BABAM1
Gene Symbol BABAM1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EEEHSAEPRPRTRSNPEGAEDRAVGAQASVGSRSEGEGEAASADDGSLNTSGAGPKSWQVPPPAPEVQIRTPRVNCPEKVIICL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.