missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAALC Partial ORF (NP_079088.1, 61 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079870-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005655

  • £280.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene was identified by gene expression studies in patients with acute myeloid leukemia (AML). The gene is conserved among mammals and is not found in lower organisms. Tissues that express this gene develop from the neuroectoderm. Multiple alternatively spliced transcript variants that encode different proteins have been described for this gene; however, some of the transcript variants are found only in AML cell lines. [provided by RefSeq]

Sequence: PSNGVPRSTAPGGIPNPEKKTNCETQCPNPQSLSSGPLTQKQNGLQTTEAKRDAKRMPAKEVTINVTDSIQQMDRSRRITKNCVN
Specifications

Specifications

NP_079088.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PSNGVPRSTAPGGIPNPEKKTNCETQCPNPQSLSSGPLTQKQNGLQTTEAKRDAKRMPAKEVTINVTDSIQQMDRSRRITKNCVN
RUO
BAALC
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79870
BAALC (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12015
BAALC
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers