missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H7/HHLA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £481.00
Specifications
| Antigen | B7-H7/HHLA2 |
|---|---|
| Clone | 907812R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Applications | Flow Cytometry, Blocking Assay, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710997
![]() |
R&D Systems
MAB80841R-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730067
![]() |
R&D Systems
MAB80841R-100 |
100 μg |
£481.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| B7-H7/HHLA2 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. |
| 907812R | |
| Flow Cytometry, Blocking Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UM44 | |
| 11148 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title