missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H6 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB71443-SP
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| B7-H6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1 | |
| Human embryonic kidney cell, HEK293-derived human B7-H6 protein Asp25-Ser262 Accession # Q68D85 | |
| 25 μg | |
| 374383 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| CyTOF, Flow Cytometry | |
| 2167B | |
| Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| Q68D85 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human B7-H6 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction