missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human B7-H3 Quantikine ELISA Kit

Product Code. p-7224931
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
Unit Size:
1 set
Pack of 96
This item is not returnable. View return policy

Product Code. 18732074

Brand: R&D Systems™ DB7H30

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human B7-H3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human B7-H3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human B7-H3/CD276 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.274 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Plasma (EDTA, Heparin), Serum, Serum, Urine, Urine
Specificity Natural and Recombinant Human B7-H3
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Gene Symbol CD276
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule
Target B7-H3/CD276
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.