missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£170.00 - £1785.00
Specifications
| Antigen | B7-H2/ICOSLG |
|---|---|
| Clone | 136726 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Neutralization 1-4 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709417
![]() |
R&D Systems
MAB165-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789408
![]() |
R&D Systems
MAB165-100 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742407
![]() |
R&D Systems
MAB165-500 |
500 μg |
£1785.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| B7-H2/ICOSLG | |
| Flow Cytometry 2.5 ug/10^6 cells, Neutralization 1-4 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| O75144 | |
| 23308 | |
| Mouse myeloma cell line NS0-derived recombinant human B7-H2 Asp19-Ser258 Accession # O75144 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human B7-H2 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) B7-1, rhB7-2, rhB7-H1, rhB7-H3, or recombinant mouse B7-H2 is observed. |
| 136726 | |
| Flow Cytometry, Neutralization, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| ICOSLG | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title