missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-2/CD86 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF-141-SP
This item is not returnable.
View return policy
Description
B7-2/CD86 Polyclonal specifically detects B7-2/CD86 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Knockout Validated.
Specifications
| B7-2/CD86 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 942 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.25-1.25 ug/mL, Knockout Validated | |
| P42081 | |
| CD86 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86 Ala23-His244 Accession # P42081 | |
| 25 μg | |
| Primary | |
| Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse B7-2 and recombinant rat B7-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction