missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Axl Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human Axl Quantikine ELISA Kit quantifies human Axl in cell culture supernates (50 μL), cell lysates (50 μL), serum (10 μL), edta plasma (10 μL), heparin plasma (10 μL), saliva (50 μL), urine (10 μL), cerebrospinal fluid (10 μL).
Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL |
| Assay Sensitivity | 4.6 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Cerebrospinal Fluid, Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human Axl |
| Gene ID (Entrez) | 558 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50 μL),Cell Lysates (50 μL),Serum (10 μL),EDTA Plasma (10 μL),Heparin Plasma (10 μL),Saliva (50 μL),Urine (10 μL),Cerebrospinal Fluid (10 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction