missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Axl Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
Brand: R&D Systems MAB154-SP
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Knockout Validated, ELISA Capture (Matched Antibody Pair).
Specifications
| Axl | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAA61243 | |
| AXL | |
| Mouse myeloma cell line NS0-derived recombinant human Axl Met1-Pro440 Accession # AAA61243 | |
| 25 μg | |
| Primary | |
| Detects human Axl in direct ELISAs. In direct ELISAs, this antibody shows no cross-reactivity with recombinant mouse Axl, recombinant human (rh) Dtk or rhMer. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF, KnockDown, ELISA | |
| 108724 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 558 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction