missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AXIN2 Partial ORF (NP_004646, 745 a.a. - 843 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008313-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16004995

  • £425.00 / 25µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Axin-related protein, Axin2, presumably plays an important role in the regulation of the stability of beta-catenin in the Wnt signaling pathway, like its rodent homologs, mouse conductin/rat axil. In mouse, conductin organizes a multiprotein complex of APC (adenomatous polyposis of the colon), beta-catenin, glycogen synthase kinase 3-beta, and conductin, which leads to the degradation of beta-catenin. Apparently, the deregulation of beta-catenin is an important event in the genesis of a number of malignancies. The AXIN2 gene has been mapped to 17q23-q24, a region that shows frequent loss of heterozygosity in breast cancer, neuroblastoma, and other tumors. Mutations in this gene have been associated with colorectal cancer with defective mismatch repair. [provided by RefSeq]

Sequence: EDHKEPKKLAGVHALQASELVVTYFFCGEEIPYRRMLKAQSLTLGHFKEQLSKKGNYRYYFKKASDEFACGAVFEEIWEDETVLPMYEGRILGKVERID
Specifications

Specifications

NP_004646
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EDHKEPKKLAGVHALQASELVVTYFFCGEEIPYRRMLKAQSLTLGHFKEQLSKKGNYRYYFKKASDEFACGAVFEEIWEDETVLPMYEGRILGKVERID
RUO
AXIN2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8313
AXIN2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AXIL/DKFZp781B0869/MGC10366/MGC126582
AXIN2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers