missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATRX (aa 2273-2413) Control Fragment Recombinant Protein

Product Code. 30205085
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205085

Brand: Invitrogen™ RP92134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51656 (PA5-51656. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ATRX encodes a protein that contains an ATPase/helicase domain, and thus it belongs to the SWI/SNF family of chromatin remodeling proteins. The mutations of this gene are associated with an X-linked mental retardation (XLMR) syndrome most often accompanied by alpha-thalassemia (ATRX) syndrome. These mutations have been shown to cause diverse changes in the pattern of DNA methylation, which may provide a link between chromatin remodeling, DNA methylation, and gene expression in developmental processes. This protein is found to undergo cell cycle-dependent phosphorylation, which regulates its nuclear matrix and chromatin association, and suggests its involvement in the gene regulation at interphase and chromosomal segregation in mitosis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P46100
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 546
Name Human ATRX (aa 2273-2413) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4833408C14Rik; AI447451; alpha thalassemia/mental retardation syndrome (X-linked); alpha thalassemia/mental retardation syndrome X-linked; alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); alpha thalassemia/mental retardation syndrome X-linked homolog; alpha thalassemia/mental retardation syndrome X-linked homolog (human); ATP-dependent helicase ATRX; ATR2; ATRX; ATRX, chromatin remodeler; DNA dependent ATPase and helicase; DXHXS6677E; helicase 2; helicase 2, X-linked; helicase II; heterochromatin protein 2; HP1 alpha-interacting protein; Hp1bp2; Hp1bp38; HP1-BP38; HP1-BP38 protein; JMS; mental retardation, X-linked 52; MRX52; MRXHF1; MRXS3; pABP-2; Rad54; RAD54L; RP5-875J14.1; SFM1; SHS; transcriptional regulator ATRX; XH2; X-linked helicase II; X-linked nuclear protein; XNP; Zinc finger helicase; Znf-HX
Common Name ATRX
Gene Symbol ATRX
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AAWAEYEAEKKGLTMRFNIPTGTNLPPVSFNSQTPYIPFNLGALSAMSNQQLEDLINQGREKVVEATNSVTAVRIQPLEDIISAVWKENMNLSEAQVQALALSRQASQELDVKRREAIYNDVLTKQQMLISCVQRILMNRR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.