missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ATP8A1 (aa 1090-1151) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62707 (PA5-62707. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The P-type adenosinetriphosphatases (P-type ATPases) are a family of proteins which use the free energy of ATP hydrolysis to drive uphill transport of ions across membranes. Several subfamilies of P-type ATPases have been identified. One subfamily catalyzes transport of heavy metal ions. Another subfamily transports non-heavy metal ions (NMHI). The protein encoded by this gene is a member of the third subfamily of P-type ATPases and acts to transport amphipaths, such as phosphatidylserine. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | Q9Y2Q0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 10396 |
| Name | Human ATP8A1 (aa 1090-1151) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI481521; AI853962; aminophospholipid translocase; APLT; Atp3a2; Atp8a1; ATPase 8A1, aminophospholipid transporter (APLT), class I; ATPase 8A1, p type; ATPase class I type 8 A member 1; ATPase II; ATPase phospholipid transporting 8A1; ATPase, aminophospholipid transporter (APLT), class I, type 8 A, member 1; ATPASEII; Atpc1; ATPIA; ATPP2; AW743152; AW822227; B230107D19Rik; Chromaffin granule ATPase II; ClassI; P4-ATPase flippase complex alpha subunit ATP8A1; phospholipid-transporting ATPase IA; probable phospholipid-transporting ATPase IA |
| Common Name | ATP8A1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction