missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATP6V1B1 (aa 1-66) Control Fragment Recombinant Protein

Product Code. 30213206
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213206

Brand: Invitrogen™ RP96039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56878 (PA5-56878. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c' and c') make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-ATPase V1 B subunit exists as two isoforms. In the inner ear, the V-ATPase B1 isoform functions in proton secretion and is required to maintain proper endolymph pH and normal auditory function. The gene encoding the human V-ATPase B1 isoform maps to chromosome 2cen-q13. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. The V-ATPase B2 isoform is expressed in kidney and is the only B isoform expressed in osteoclasts.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15313
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 525
Name Human ATP6V1B1 (aa 1-66) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ATP6B1; ATP6V1B1; ATPase H+ transporting V1 subunit B1; ATPase, H transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 1; ATPase, H+ transporting, lysosomal 56/58 kDa, V1 subunit B, isoform 1; ATPase, H+ transporting, lysosomal 56/58 kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58 kDa, V1 subunit B1 (Renal tubular acidosis with deafness); ATPase, H+ transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, V1 subunit B, isoform 1; AW208839; D630003L15; D630030L16Rik; D630039P21Rik; endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58 kD subunit; H+-ATPase beta 1 subunit; lysosomal 56/58 kDa; RTA1B; vacuolar H+-ATPase; vacuolar proton pump 3; vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3; VATB; V-ATPase B1; V-ATPase B1 subunit; V-ATPase subunit B 1; VMA2; VPP3; Vpp-3; V-type proton ATPase subunit B, kidney isoform
Common Name V-ATPase B1
Gene Symbol Atp6v1b1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.