missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATP6AP1L (aa 74-148) Control Fragment Recombinant Protein

Product Code. 30195687
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195687

Brand: Invitrogen™ RP96985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62837 (PA5-62837. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ATP6AP1L gene ontology annotations related to this gene include ATP hydrolysis coupled proton transport.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q52LC2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 92270
Name Human ATP6AP1L (aa 74-148) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ATP6AP1L; ATPase H+ transporting accessory protein 1 like; ATPase, H+ transporting, lysosomal accessory protein 1-like; Vacuolar proton pump subunit S1-like protein; V-type proton ATPase subunit S1-like protein
Common Name ATP6AP1L
Gene Symbol ATP6AP1L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AVFNPTGVYAPSGYSYRCQRVGSLQQDQALLLPSDTDDGSSLWEVTFIDFQIQGFAIKGGRFTKAQDCASSFSPA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.