missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATP5J2 (aa 75-94) Control Fragment Recombinant Protein

Product Code. 30213170
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213170

Brand: Invitrogen™ RP105503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67104 (PA5-67104. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ATP5J2 is part of the complex F0 domain of the mitochondrial ATP synthase. This produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. It is composed of two complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, which comprises the proton channel. ATP5J2 encodes the f subunit of the Fo complex. There are 2 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P56134
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9551
Name Human ATP5J2 (aa 75-94) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110019H14Rik; ATP synthase f chain, mitochondrial; ATP synthase membrane subunit f; ATP synthase subunit f, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit f, isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2; ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2; ATP5J2; ATP5JL; Atp5mf; F1F0-type ATPase subunit f; F1Fo-ATP synthase complex Fo membrane domain f subunit; F1Fo-ATPase synthase f subunit; hypothetical protein, clone:2-31
Common Name ATP5J2
Gene Symbol ATP5MF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LFSYSFSYKHLKHERLRKYH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.