missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATP2C2 (aa 491-547) Control Fragment Recombinant Protein

Product Code. 30206059
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206059

Brand: Invitrogen™ RP108060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67396 (PA5-67396. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ATP2C2, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 2, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in the related gene ATP2C1 cause Hailey-Hailey disease, for which ATP2C2 does not compensate, suggesting that ATP2C2 plays other physiological roles. Unlike ATP2C1, ATP2C2 has a much more restricted expression pattern and displays a higher maximal turnover rate for overall Ca2+-ATPase reaction and a lower apparent affinity for cytosolic Ca2+ activation of phosphorylation. Overexpression of ATP2C2 in mammary tumors result a Ca2+ influx via the store-operated Ca2+ channel ORAI1 and independent of the STIM1 and STIM2 sensors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75185
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9914
Name Human ATP2C2 (aa 491-547) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810010G06Rik; Atp2c2; ATPase 2C2; ATPase secretory pathway Ca2+ transporting 2; ATPase, Ca++ transporting, type 2 C, member 2; calcium-transporting ATPase type 2 C member 2; Kiaa0703; mKIAA0703; Secretory pathway Ca(2+)-ATPase 2; secretory pathway Ca2+-ATPase; secretory pathway calcium ATPase 2; Spca2
Common Name ATP2C2
Gene Symbol ATP2C2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WMAVKCSLKTEDQEDIYFMKGALEEVIRYCTMYNNGGIPLPLTPQQRSFCLQEEKRM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.