missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ATG16L2 (aa 262-348) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61000 (PA5-61000. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May play a role in autophagy. [UniProt]
Specifikationer
Specifikationer
| Accession Number | Q8NAA4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 89849 |
| Name | Human ATG16L2 (aa 262-348) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2410118P20Rik; APG16-like 2; ATG16 autophagy related 16-like 2; ATG16B; Atg16l2; Atg16L2 alpha; Atg16L2 beta; autophagy related 16 like 2; autophagy related 16-like 2; autophagy related 16-like 2 (S. cerevisiae); autophagy-related 16-like 2 alpha; autophagy-related 16-like 2 beta; autophagy-related protein 16-2; mFLJ00012 protein; RGD1311400; WD repeat-containing protein 80; WDR80 |
| Common Name | ATG16L2 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering