missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AspRS (aa 360-442) Control Fragment Recombinant Protein

Product Code. 30201572
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201572

Brand: Invitrogen™ RP94127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55332 (PA5-55332. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. It is a mitochondrial enzyme that specifically aminoacylates aspartyl-tRNA. Mutations in this gene are associated with leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation (LBSL).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6PI48
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55157
Name Human AspRS (aa 360-442) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5830468K18Rik; aspartate tRNA ligase 2, mitochondrial; aspartate--tRNA ligase, mitochondrial; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase 2 (mitochondrial); aspartyl-tRNA synthetase 2, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartate--tRNA ligase, mitochondrial; aspRS; Dars2; LBSL; MT-ASPRS; RGD1308286; si:dkey-21n10.1
Common Name AspRS
Gene Symbol DARS2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSKPHGTVKAICIPEGAKYLKRKDIESIRNFAADHFNQEILPVFLNANRNWNSPVANFIMESQRLELIRLMETQEEDVVLLTA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato