missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ASPH (aa 506-600) Control Fragment Recombinant Protein

Product Code. 30207190
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207190

Brand: Invitrogen™ RP105009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65929 (PA5-65929. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins. This gene is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12797
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 444
Name Human ASPH (aa 506-600) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310005F16Rik; 3110001L23Rik; A beta H-J-J; AAH; AI848629; ASP beta-hydroxylase; Aspartate beta-hydroxylase; aspartate-beta-hydroxylase; aspartyl (asparaginyl) beta hydroxylase; aspartyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase-like; aspartyl/asparaginyl-beta-hydroxylase; ASPH; AW261690; AW561901; BAH; C79816; calsequestrin-binding protein; cardiac junctin; CASQ2BP1; cI-37; FDLAB; HAAH; humbug; JCTN; jumbug; junctate; junctin; junctional sarcoplasmic reticulum protein; peptide-aspartate beta-dioxygenase; Unknown (protein for MGC:137539)
Common Name ASPH
Gene Symbol Asph
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ESIPYLKEGIESGDPGTDDGRFYFHLGDAMQRVGNKEAYKWYELGHKRGHFASVWQRSLYNVNGLKAQPWWTPKETGYTELVKSLERNWKLIRDE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.