missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASGR1/ASGPR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4394-100
This item is not returnable.
View return policy
Description
ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| ASGPR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ASGPR, ASGPR 1, ASGP-R 1, ASGPR1, asialoglycoprotein receptor 1, CLEC4H1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1, MHL1, RHL1 | |
| Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1 Gln62-Leu291 Accession # P07306 | |
| 100 μg | |
| Primary | |
| Detects human ASGR/1ASGPR1 in direct ELISAs. | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 950216 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
| P07306 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction