missing translation for 'onlineSavingsMsg'
Learn More

Human ASGR1/ASGPR1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18743959
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18743959 25 μg 25µg
18749438 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18743959 Supplier R&D Systems Supplier No. MAB4394SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASGPR1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 950216
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P07306
Gene Alias ASGPR, ASGPR 1, ASGP-R 1, ASGPR1, asialoglycoprotein receptor 1, CLEC4H1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1, MHL1, RHL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1 Gln62-Leu291 Accession # P07306
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 432
Test Specificity Detects human ASGR/1ASGPR1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.