missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASC Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3805
This item is not returnable.
View return policy
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| ASC/TMS1 | |
| Polyclonal | |
| Unconjugated | |
| Q9ULZ3 | |
| PYCARD | |
| E. coli-derived recombinant human ASC Leu112-Ser195 Accession # Q9ULZ3 | |
| 100 μg | |
| Primary | |
| Detects human ASC in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| ASCMGC10332, CARD5, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYCARD, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29108 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction