missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ASAP3 (aa 507-568) Control Fragment Recombinant Protein

Product Code. 30204094
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204094

Brand: Invitrogen™ RP91722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110863 (PA5-110863. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TDY4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55616
Name Human ASAP3 (aa 507-568) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9430088F20Rik; ACAP4; ARF6 GTPase-activating protein; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1; Asap3; centaurin, beta 6; CENTB6; DDEFL1; development and differentiation enhancing factor-like 1; development and differentiation-enhancing factor-like 1; Gm140; Protein up-regulated in liver cancer 1; UPLC1; up-regulated in liver cancer 1 (UPLC1)
Common Name ASAP3
Gene Symbol Asap3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EAQLPSHGGPKPSAESDMGTRRDYIMAKYVEHRFARRCTPEPQRLWTAICNRDLLSVLEAFA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.