missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARS2 (aa 420-516) Control Fragment Recombinant Protein

Product Code. 30198081
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198081

Brand: Invitrogen™ RP92626

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60148 (PA5-60148. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Acts as a mediator between the cap-binding complex (CBC) and the primary microRNAs (miRNAs) processing machinery during cell proliferation. Contributes to the stability and delivery of capped primary miRNA transcripts to the primary miRNA processing complex containing DGCR8 and DROSHA, thereby playing a role in RNA-mediated gene silencing (RNAi) by miRNAs. Binds capped RNAs (m7GpppG-capped RNA); however interaction is probably mediated via its interaction with NCBP1/CBP80 component of the CBC complex. Involved in cell cycle progression at S phase. Does not directly confer arsenite resistance but rather modulates arsenic sensitivity. Independently of its activity on miRNAs, necessary and sufficient to promote neural stem cell self-renewal. Does so by directly binding SOX2 promoter and positively regulating its transcription. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXP5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51593
Name Human ARS2 (aa 420-516) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2810019G02Rik; Ars2; arsenate resistance protein 2; arsenate resistance protein ARS2; arsenite resistance protein; arsenite resistance protein 2; arsenite-resistance protein 2; Asr2; ASR2A; ASR2B; ASR2C; ASR2D; ch211-283g2.4; CHUNP6881; LOC686980; serrate; Serrate RNA effector molecule homolog; serrate RNA effector molecule homolog (Arabidopsis); serrate, RNA effector molecule; similar to arsenate resistance protein 2; srrt; zgc:92060
Common Name ARS2
Gene Symbol SRRT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TCSLFMRNIAPNISRAEIISLCKRYPGFMRVALSEPQPERRFFRRGWVTFDRSVNIKEICWNLQNIRLRECELSPGVNRDLTRRVRNINGITQHKQI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.