missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARPC5 Partial ORF (AAH57237.1, 31 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010092-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005215

  • £280.00 / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined. [provided by RefSeq]

Sequence: GDGQAGPDEGEVDSCLRHSITGNMTAALQAALKNPPINTKSQAVKDRAGSIVLKVLISF
Specifications

Specifications

AAH57237.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.23kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GDGQAGPDEGEVDSCLRHSITGNMTAALQAALKNPPINTKSQAVKDRAGSIVLKVLISF
RUO
ARPC5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10092
ARPC5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARC16/MGC88523/dJ127C7.3/p16-Arc
ARPC5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers