missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ARNT/HIF-1 beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5630-SP
This item is not returnable.
View return policy
Description
ARNT/HIF-1 beta Monoclonal specifically detects ARNT/HIF-1 beta in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| ARNT/HIF-1 beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P27540 | |
| ARNT | |
| E. coli-derived recombinant human ARNT/HIF-1 beta Asp517-Ala691 Accession # P27540 | |
| 25 μg | |
| Primary | |
| Detects human ARNT/HIF-1 beta in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 775146 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF1 beta, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 405 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction