missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARMER (aa 92-132) Control Fragment Recombinant Protein

Product Code. 30203246
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203246

Brand: Invitrogen™ RP91178

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60977 (PA5-60977. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Apoptosis is important for normal development and tissue homeostasis. It is mediated by various caspases and ultimately results in the activation of endogenous endonucleases that degrade cellular DNA. Although apoptosis induced by endoplasmic reticulum (ER) stress is thought to be mediated by caspase-12, other caspases such as caspase-9 are also thought to be activated following ER stress. Recently, ARMER, a novel integral ER-membrane protein was shown to protect cells from ER stress-induced apoptosis. Analysis of the caspase proteolytic cascade suggests that ARMER acts by inhibiting caspase-9 activity, although the mechanism for this remains unknown. It should be noted that ARMER is not related to the inhibitor of apoptosis proteins (IAP) family and does not contain any baculoviral IAP repeat (BIR) domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15041
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23204
Name Human ARMER (aa 92-132) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ADP ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor GTPase 6 interacting protein 1; ADP-ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor-like 6 interacting protein; ADP-ribosylation factor-like 6 interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1-like protein; ADP-ribosylation-like factor 6 interacting protein; ADP-ribosylation-like factor 6-interacting protein; AIP1; aip-1; AIP-6; AL022945; apoptotic regulator in the membrane of the endoplasmic reticulum; ARL-6-interacting protein 1; Arl6ip; arl6ip protein; Arl6ip1; ARMER; AU042858; C85138; KIAA0069; mKIAA0069; Protein TB x 2; SPG61; zgc:56593; zgc:77873
Common Name ARMER
Gene Symbol ARL6IP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FGSNKWTTEQQQRFHEICSNLVKTRRRAVGWWKRLFTLKEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.